missing translation for 'onlineSavingsMsg'
Learn More

Ethyl iodoacetate, 98%, stabilized, Thermo Scientific Chemicals

$572.07 - $572.07

Chemical Identifiers

CAS 623-48-3
Molecular Formula C4H7IO2
Molecular Weight (g/mol) 214.00
MDL Number MFCD00001081
InChI Key MFFXVVHUKRKXCI-UHFFFAOYSA-N
Synonym ethyl iodoacetate, acetic acid, iodo-, ethyl ester, ethyl monoiodoacetate, iodoacetic acid, ethyl ester, acetic acid, 2-iodo-, ethyl ester, ethylester kyseliny jodoctove czech, acetic acid, 2-iodo-,ethyl ester, ethylester kyseliny jodoctove, ethyliodoacetate, iodoacetic acid ethyl
PubChem CID 12183
IUPAC Name ethyl 2-iodoacetate
SMILES CCOC(=O)CI
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC222190500
SDS View Documents
Thermo Scientific Chemicals
222190500
50 mL Glass bottle
Each for $572.07
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

623-48-3
214.00
MFFXVVHUKRKXCI-UHFFFAOYSA-N
12183
CCOC(=O)CI
C4H7IO2
MFCD00001081
ethyl iodoacetate, acetic acid, iodo-, ethyl ester, ethyl monoiodoacetate, iodoacetic acid, ethyl ester, acetic acid, 2-iodo-, ethyl ester, ethylester kyseliny jodoctove czech, acetic acid, 2-iodo-,ethyl ester, ethylester kyseliny jodoctove, ethyliodoacetate, iodoacetic acid ethyl
ethyl 2-iodoacetate

Specifications

623-48-3
1.8000g/mL
76°C
97.5% min. (GC)
C4H7IO2
ICH2CO2C2H5
50 mL
1.8
Solubility in water: immiscible
CCOC(=O)CI
214.00
12183
98%
Ethyl iodoacetate, 98%
Orange to Yellow
179°C to 180°C
Authentic
Glass bottle
1.5035 to 1.5055
MFCD00001081
02,222
ethyl iodoacetate, acetic acid, iodo-, ethyl ester, ethyl monoiodoacetate, iodoacetic acid, ethyl ester, acetic acid, 2-iodo-, ethyl ester, ethylester kyseliny jodoctove czech, acetic acid, 2-iodo-,ethyl ester, ethylester kyseliny jodoctove, ethyliodoacetate, iodoacetic acid ethyl
MFFXVVHUKRKXCI-UHFFFAOYSA-N
ethyl 2-iodoacetate
copper
214
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Toxic if swallowed.
Toxic if inhaled.
Toxic in contact with skin.
Causes skin irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 210-796-3

RTECSNumber : AI3575000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only