missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Indazole-5-carboxylate 98.0+%, TCI America™

Catalog No. E10675G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10675G 5 g
E10671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10675G Supplier TCI America Supplier No. E10675G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 192944-51-7
Molecular Formula C10H10N2O2
Molecular Weight (g/mol) 190.20
MDL Number MFCD03426220
InChI Key SKABXDPLIJIWLR-UHFFFAOYSA-N
Synonym 1h-indazole-5-carboxylic acid ethyl ester, 1h-indazole-5-carboxylic acid, ethyl ester, 5-ethoxycarbonyl-1h-indazole, 1h-indazole-5-carboxylicacidethylester, pubchem18041, 5-ethoxycarbonylindazole, acmc-1c4yw, ethyl indazole-5-carboxylate, ethyl 2h-indazole-5-carboxylate, indazole-5-carboxylic acid ethyl ester
PubChem CID 1501980
IUPAC Name ethyl 1H-indazole-5-carboxylate
SMILES CCOC(=O)C1=CC=C2NN=CC2=C1

Specifications

Melting Point 138°C
Color Yellow
Quantity 5 g
Formula Weight 190.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl Indazole-5-carboxylate