CAS RN 192944-51-7
IUPAC Name:
ethyl 1H-indazole-5-carboxylate
Synonyms:
ethyl 1H-indazole-5-carboxylate
Molecular Weight (g/mol):
190.2
Molecular Formula:
C10H10N2O2
InChi Key:
SKABXDPLIJIWLR-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CC=C2NN=CC2=C1
1
–
1
of
1
results
1
Ethyl Indazole-5-carboxylate 98.0+%, TCI America™
CAS: 192944-51-7 Molecular Formula: C10H10N2O2 Molecular Weight (g/mol): 190.20 MDL Number: MFCD03426220 InChI Key: SKABXDPLIJIWLR-UHFFFAOYSA-N Synonym: 1h-indazole-5-carboxylic acid ethyl ester,1h-indazole-5-carboxylic acid, ethyl ester,5-ethoxycarbonyl-1h-indazole,1h-indazole-5-carboxylicacidethylester,pubchem18041,5-ethoxycarbonylindazole,acmc-1c4yw,ethyl indazole-5-carboxylate,ethyl 2h-indazole-5-carboxylate,indazole-5-carboxylic acid ethyl ester PubChem CID: 1501980 IUPAC Name: ethyl 1H-indazole-5-carboxylate SMILES: CCOC(=O)C1=CC=C2NN=CC2=C1