missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl hydrogen suberate, 98%
CAS: 14113-01-0 | C10H18O4 | 202.25 g/mol
$157.73 - $590.18
Chemical Identifiers
| CAS | 14113-01-0 |
|---|---|
| Molecular Formula | C10H18O4 |
| Molecular Weight (g/mol) | 202.25 |
| MDL Number | MFCD00014385 |
| InChI Key | KCFVQEAPUOVXTH-UHFFFAOYSA-N |
| PubChem CID | 84204 |
| IUPAC Name | 8-ethoxy-8-oxooctanoic acid |
| SMILES | CCOC(=O)CCCCCCC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAL0427206
|
N/A | 5 g |
Each for $157.73
|
|
|||||
|
AAL0427214
|
N/A | 25 g |
Each for $590.18
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 14113-01-0 | |
| 202.25 | |
| KCFVQEAPUOVXTH-UHFFFAOYSA-N | |
| 8-ethoxy-8-oxooctanoic acid |
| C10H18O4 | |
| MFCD00014385 | |
| 84204 | |
| CCOC(=O)CCCCCCC(=O)O |
Specifications
| 14113-01-0 | |
| C10H18O4 | |
| 1775737 | |
| KCFVQEAPUOVXTH-UHFFFAOYSA-N | |
| 8-ethoxy-8-oxooctanoic acid | |
| 84204 | |
| 98% |
| 25°C to 27°C | |
| MFCD00014385 | |
| ethyl hydrogen suberate, octanedioic acid, ethyl, korksaureathylester, acmc-1b2e5, 8-ethoxy-8-oxo-octanoic acid, octanedioic acid 1-ethyl ester, octanedioic acid,1-ethyl ester, octanedioic acid, 1-ethyl ester, 8-ethoxy-8-oxidanylidene-octanoic acid, ethylhydrogensuberate, monoethylsuberate; subericacid monoethylester | |
| CCOC(=O)CCCCCCC(=O)O | |
| 202.25 | |
| 202.25 | |
| Ethyl hydrogen suberate |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 237-968-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only