missing translation for 'onlineSavingsMsg'
Learn More

Ethyl hydrogen suberate, 98%

Catalog No. AAL0427214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0427214 25 g
AAL0427206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0427214 Supplier Thermo Scientific Chemicals Supplier No. L0427214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14113-01-0 | C10H18O4 | 202.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14113-01-0
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00014385
InChI Key KCFVQEAPUOVXTH-UHFFFAOYSA-N
PubChem CID 84204
IUPAC Name 8-ethoxy-8-oxooctanoic acid
SMILES CCOC(=O)CCCCCCC(=O)O

Specifications

Melting Point 25°C to 27°C
Quantity 25 g
Beilstein 1775737
Synonym ethyl hydrogen suberate, octanedioic acid, ethyl, korksaureathylester, acmc-1b2e5, 8-ethoxy-8-oxo-octanoic acid, octanedioic acid 1-ethyl ester, octanedioic acid,1-ethyl ester, octanedioic acid, 1-ethyl ester, 8-ethoxy-8-oxidanylidene-octanoic acid, ethylhydrogensuberate, monoethylsuberate; subericacid monoethylester
Formula Weight 202.25
Percent Purity 98%
Chemical Name or Material Ethyl hydrogen suberate
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 237-968-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only