missing translation for 'onlineSavingsMsg'
Learn More

Ethyl hydrogen sebacate, 98%

Catalog No. p-7044027
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0413406 5 g
AAL0413414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0413406 Supplier Thermo Scientific Chemicals Supplier No. L0413406

CAS: 693-55-0 | C12H22O4 | 230.304 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 693-55-0
Molecular Formula C12H22O4
Molecular Weight (g/mol) 230.304
MDL Number MFCD00014387
InChI Key DLZCDMPHHUODDO-UHFFFAOYSA-N
Synonym ethyl hydrogen sebacate, monoethyl sebacate, sebacic acid, monoethyl ester, decanedioic acid monoethyl ester, decanedioic acid, 1-ethyl ester, decanedioic acid, monoethyl ester, sebacinsaureathylester, mono-ethyl sebacate, sebacic acid 1-ethyl ester, decanedioic acid 1-ethyl ester
PubChem CID 69660
IUPAC Name 10-ethoxy-10-oxodecanoic acid
SMILES CCOC(=O)CCCCCCCCC(=O)O

Specifications

Melting Point 35°C to 38°C
Quantity 5 g
Beilstein 1782569
Formula Weight 230.3
Percent Purity 98%
Chemical Name or Material Ethyl hydrogen sebacate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-753-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only