CAS RN 693-55-0
IUPAC Name:
10-ethoxy-10-oxodecanoic acid
Synonyms:
10-ethoxy-10-oxodecanoic acid
Molecular Weight (g/mol):
230.3
Molecular Formula:
C12H22O4
InChi Key:
DLZCDMPHHUODDO-UHFFFAOYSA-N
SMILES:
CCOC(=O)CCCCCCCCC(O)=O
1
–
1
of
1
results
1
Ethyl hydrogen sebacate, 98%
CAS: 693-55-0 Molecular Formula: C12H22O4 Molecular Weight (g/mol): 230.304 MDL Number: MFCD00014387 InChI Key: DLZCDMPHHUODDO-UHFFFAOYSA-N Synonym: ethyl hydrogen sebacate,monoethyl sebacate,sebacic acid, monoethyl ester,decanedioic acid monoethyl ester,decanedioic acid, 1-ethyl ester,decanedioic acid, monoethyl ester,sebacinsaureathylester,mono-ethyl sebacate,sebacic acid 1-ethyl ester,decanedioic acid 1-ethyl ester PubChem CID: 69660 IUPAC Name: 10-ethoxy-10-oxodecanoic acid SMILES: CCOC(=O)CCCCCCCCC(=O)O