missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Hydrogen Maleate 95.0+%, TCI America™
Supplier: TCI America M0614100ML
Specifications
| Ethyl Hydrogen Maleate | |
| Yellow | |
| MFCD00063175 | |
| Maleic Acid Monoethyl Ester, Monoethyl Maleate | |
| CCOC(=O)C=CC(=O)O | |
| 144.13 | |
| 144.13 | |
| Liquid |
| 3990-03-2 | |
| C6H8O4 | |
| 100 mL | |
| XLYMOEINVGRTEX-ARJAWSKDSA-N | |
| (Z)-4-ethoxy-4-oxobut-2-enoic acid | |
| 5354457 | |
| ≥95.0% (T) |
Chemical Identifiers
| 3990-03-2 | |
| 144.13 | |
| XLYMOEINVGRTEX-ARJAWSKDSA-N | |
| 5354457 | |
| CCOC(=O)C=CC(=O)O |
| C6H8O4 | |
| MFCD00063175 | |
| Maleic Acid Monoethyl Ester, Monoethyl Maleate | |
| (Z)-4-ethoxy-4-oxobut-2-enoic acid |
Safety and Handling
EINECSNumber : (2)-1105&(2)-1142
RTECSNumber : EM6538750
TSCA : Yes
Recommended Storage : Refrigerator