accessibility menu, dialog, popup

UserName

CAS RN 3990-03-2

CH3OOOHO(Z)

CAS RN 3990-03-2

IUPAC Name: (2Z)-4-ethoxy-4-oxobut-2-enoic acid
Synonyms: (2Z)-4-ethoxy-4-oxobut-2-enoic acid
Molecular Weight (g/mol): 144.13
Molecular Formula: C6H8O4
InChi Key: XLYMOEINVGRTEX-ARJAWSKDSA-N
SMILES: CCOC(=O)\C=C/C(O)=O
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results