missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Hydrogen Maleate 95.0+%, TCI America™

Catalog No. M061425ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M061425ML 25 mL
M0614100ML 100 mL
M0614500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M061425ML Supplier TCI America Supplier No. M061425ML
Only null left

Chemical Identifiers

CAS 3990-03-2
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.13
MDL Number MFCD00063175
InChI Key XLYMOEINVGRTEX-ARJAWSKDSA-N
Synonym Maleic Acid Monoethyl Ester, Monoethyl Maleate
PubChem CID 5354457
IUPAC Name (Z)-4-ethoxy-4-oxobut-2-enoic acid
SMILES CCOC(=O)C=CC(=O)O

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 144.13
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material Ethyl Hydrogen Maleate
EINECSNumber (2)-1105&(2)-1142
RTECSNumber EM6538750
TSCA Yes
Recommended Storage Refrigerator