missing translation for 'onlineSavingsMsg'
Learn More

Ethyl hippurate, 96%

Catalog No. AAB2476514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2476514 25 g
AAB2476506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2476514 Supplier Thermo Scientific Chemicals Supplier No. B2476514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1499-53-2 | C11H13NO3 | 207.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1499-53-2
Molecular Formula C11H13NO3
Molecular Weight (g/mol) 207.23
MDL Number MFCD00026890
InChI Key PTXRQIPIELXJFH-UHFFFAOYSA-N
Synonym ethyl hippurate, ethyl n-benzoylglycinate, n-benzoylglycine ethyl ester, ethyl 2-phenylformamido acetate, benzoic amide, n-ethoxycarbonyl methyl, ethylhippurate, ethyl benzoylaminoacetate, acmc-1buir, hippuric acid ethyl ester, maybridge1_006087
PubChem CID 226558
IUPAC Name ethyl 2-benzamidoacetate
SMILES CCOC(=O)CNC(=O)C1=CC=CC=C1

Specifications

Melting Point 58°C to 60°C
Quantity 25 g
Formula Weight 207.23
Percent Purity 98%
Chemical Name or Material Ethyl hippurate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only