missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Hexanoate 99.0+%, TCI America™
$31.68 - $108.11
Chemical Identifiers
| CAS | 123-66-0 |
|---|---|
| Molecular Formula | C8H16O2 |
| Molecular Weight (g/mol) | 144.214 |
| MDL Number | MFCD00009511 |
| InChI Key | SHZIWNPUGXLXDT-UHFFFAOYSA-N |
| Synonym | ethyl caproate, ethyl hexoate, hexanoic acid, ethyl ester, hexanoic acid ethyl ester, capronic ether absolute, caproic acid ethyl ester, ethyl n-hexanoate, ethyl butyl acetate, caproate, acetic acid, butyl-, ethyl ester |
| PubChem CID | 31265 |
| ChEBI | CHEBI:86055 |
| IUPAC Name | ethyl hexanoate |
| SMILES | CCCCCC(=O)OCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
H010825ML
|
TCI America
H010825ML |
25 mL |
Each for $31.68
|
|
|||||
|
H0108500ML
|
TCI America
H0108500ML |
500 mL |
Each for $108.11
|
|
|||||
Chemical Identifiers
| 123-66-0 | |
| 144.214 | |
| SHZIWNPUGXLXDT-UHFFFAOYSA-N | |
| 31265 | |
| ethyl hexanoate |
| C8H16O2 | |
| MFCD00009511 | |
| ethyl caproate, ethyl hexoate, hexanoic acid, ethyl ester, hexanoic acid ethyl ester, capronic ether absolute, caproic acid ethyl ester, ethyl n-hexanoate, ethyl butyl acetate, caproate, acetic acid, butyl-, ethyl ester | |
| CHEBI:86055 | |
| CCCCCC(=O)OCC |
Specifications
| 123-66-0 | |
| 167°C | |
| MFCD00009511 | |
| 3272 | |
| SHZIWNPUGXLXDT-UHFFFAOYSA-N | |
| ethyl hexanoate | |
| 31265 | |
| 144.21 | |
| Liquid |
| Colorless | |
| C8H16O2 | |
| 25 mL | |
| ethyl caproate, ethyl hexoate, hexanoic acid, ethyl ester, hexanoic acid ethyl ester, capronic ether absolute, caproic acid ethyl ester, ethyl n-hexanoate, ethyl butyl acetate, caproate, acetic acid, butyl-, ethyl ester | |
| CCCCCC(=O)OCC | |
| 144.214 | |
| CHEBI:86055 | |
| ≥99.0% (GC) | |
| Ethyl Hexanoate |
Safety and Handling
EINECSNumber : (2)-0782
RTECSNumber : MO7735000
TSCA : Yes