missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Hexanoate 99.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 167°C |
| Quantity | 25 mL |
| UN Number | 3272 |
| Formula Weight | 144.21 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Ethyl Hexanoate |
Safety and Handling
| EINECSNumber | (2)-0782 |
| RTECSNumber | MO7735000 |
| TSCA | Yes |