missing translation for 'onlineSavingsMsg'
Learn More

Ethyl heptanoate, Analytical Standard, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7193480
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019725 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019725 Supplier Merck Emd Millipore Supplier No. 975871ML
Only null left

Specifications

CAS 106-30-9
Density 0.87 g/mL (at 25°C)
Physical Form Neat
Linear Formula CH3(CH2)5 COOC2H5
MDL Number MFCD00009538
Melting Point -66°C
Boiling Point 188°C to 189°C
Synonym Ethyl enanthate
Quantity 1 mL
Formula Weight 158.24
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less