missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Heptadecafluorononanoate 98.0+%, TCI America™

Numéro de catalogue. H10385G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
H10385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. H10385G Fournisseur TCI America Code fournisseur H10385G
Il en reste null

Identifiants chimiques

CAS 30377-52-7
Molecular Formula C11H5F17O2
Molecular Weight (g/mol) 492.13
MDL Number MFCD00039245
InChI Key DRLDSHOYANTUND-UHFFFAOYSA-N
Synonym Heptadecafluorononanoic Acid Ethyl Ester, Ethyl Perfluorononanoate, Perfluorononanoic Acid Ethyl Ester
PubChem CID 121681
IUPAC Name ethyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluorononanoate
SMILES CCOC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Spécifications

Color Colorless
Quantity 5 g
Formula Weight 492.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Heptadecafluorononanoate