missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Glyoxylate Polymer form (47% in Toluene), TCI America™

Catalog No. G026425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G026425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G026425G Supplier TCI America Supplier No. G026425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 924-44-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.089
MDL Number MFCD00044009
InChI Key DBPFRRFGLYGEJI-UHFFFAOYSA-N
Synonym ethyl glyoxalate, ethyl glyoxylate, ethyl oxoacetate, glyoxylic acid ethyl ester, acetic acid, oxo-, ethyl ester, ethyl glyoxalate solution, unii-ixx9mq1j1i, acetic acid, 2-oxo-, ethyl ester, ixx9mq1j1i, 2-oxoacetic acid, ethyl ester
PubChem CID 70211
ChEBI CHEBI:53275
IUPAC Name ethyl 2-oxoacetate
SMILES CCOC(=O)C=O

Specifications

Boiling Point 110°C
Quantity 25 g
UN Number 1294
Physical Form Liquid
Chemical Name or Material Ethyl Glyoxylate Polymer form (47% in Toluene)