missing translation for 'onlineSavingsMsg'
Learn More

Ethyl glyoxylate, ca 50% soln. in toluene

Catalog No. p-7047427
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1920722 100 g
AAL1920736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1920722 Supplier Thermo Scientific Chemicals Supplier No. L1920722

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 924-44-7 | C4H6O3 | 102.089 g/mol

Ethyl glyoxylate is widely used as an intermediate in the synthesis of pharmaceuticals (high reactivity of aldehyde function). It is used in the synthesis of a biodegradable polymer, poly(ethyl glyoxylate). It is actively involved in the Friedel-Crafts alkylation reactions with thiophenes to give the corresponding secondary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl glyoxylate is widely used as an intermediate in the synthesis of pharmaceuticals (high reactivity of aldehyde function). It is used in the synthesis of a biodegradable polymer, poly(ethyl glyoxylate). It is actively involved in the Friedel-Crafts alkylation reactions with thiophenes to give the corresponding secondary alcohols.

Solubility
Immiscible with water.

Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 924-44-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.089
MDL Number MFCD00044009
InChI Key DBPFRRFGLYGEJI-UHFFFAOYSA-N
Synonym ethyl glyoxalate, ethyl glyoxylate, ethyl oxoacetate, glyoxylic acid ethyl ester, acetic acid, oxo-, ethyl ester, ethyl glyoxalate solution, unii-ixx9mq1j1i, acetic acid, 2-oxo-, ethyl ester, ixx9mq1j1i, 2-oxoacetic acid, ethyl ester
PubChem CID 70211
ChEBI CHEBI:53275
IUPAC Name ethyl 2-oxoacetate
SMILES CCOC(=O)C=O

Specifications

Density 1.003
Boiling Point 110°C
Flash Point 7°C (44°F)
Refractive Index 1.475
Quantity 100 g
UN Number UN1993
Beilstein 1209486
Sensitivity Air Sensitive
Solubility Information Immiscible with water.
Formula Weight 102.09
Concentration or Composition (by Analyte or Components) ≈50% soln. in toluene
Chemical Name or Material Ethyl glyoxylate
Show More Show Less
Hazard Category H225-H304-H315-H317-H319-H335-H336-H361d-H373
Hazard Statement GHS H Statement
H225-H304-H361-H373-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P272-P280-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 213-105-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only