missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl glyoxalate, technical, ∼50% in toluene
CAS: 924-44-7 | C4H6O3 | 102.09 g/mol
Supplier: Thermo Scientific Chemicals 425760010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Ethyl glyoxalate | |
| 924-44-7 | |
| 110°C | |
| Glass bottle | |
| HCOCO2CH2CH3 | |
| 1.032 | |
| DBPFRRFGLYGEJI-UHFFFAOYSA-N | |
| ethyl 2-oxoacetate | |
| 70211 | |
| 102.09 |
| Approx. 50% in toluene | |
| 1.0320g/mL | |
| 7°C | |
| C4H6O3 | |
| 1 kg | |
| ethyl glyoxalate, ethyl glyoxylate, ethyl oxoacetate, glyoxylic acid ethyl ester, acetic acid, oxo-, ethyl ester, ethyl glyoxalate solution, unii-ixx9mq1j1i, acetic acid, 2-oxo-, ethyl ester, ixx9mq1j1i, 2-oxoacetic acid, ethyl ester | |
| CCOC(=O)C=O | |
| 102.09 | |
| CHEBI:53275 | |
| Technical |
Chemical Identifiers
| 924-44-7 | |
| 102.09 | |
| ethyl glyoxalate, ethyl glyoxylate, ethyl oxoacetate, glyoxylic acid ethyl ester, acetic acid, oxo-, ethyl ester, ethyl glyoxalate solution, unii-ixx9mq1j1i, acetic acid, 2-oxo-, ethyl ester, ixx9mq1j1i, 2-oxoacetic acid, ethyl ester | |
| CHEBI:53275 | |
| CCOC(=O)C=O |
| C4H6O3 | |
| DBPFRRFGLYGEJI-UHFFFAOYSA-N | |
| 70211 | |
| ethyl 2-oxoacetate |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 213-105-3
RUO – Research Use Only