missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Glycolate 98.0+%, TCI America™

Catalog No. p-7130205
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G024225G 25 g
G0242100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G024225G Supplier TCI America Supplier No. G024225G

Chemical Identifiers

CAS 623-50-7
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
MDL Number MFCD00021970
InChI Key ZANNOFHADGWOLI-UHFFFAOYSA-N
Synonym ethyl glycolate, ethyl hydroxyacetate, acetic acid, hydroxy-, ethyl ester, glycolic acid, ethyl ester, ethyl glycollate, hydroxyacetic acid ethyl ester, glycolic acid ethyl ester, unii-c306e91wfk, acetic acid, 2-hydroxy-, ethyl ester, 2-hydroxyacetic acid ethyl ester
PubChem CID 12184
IUPAC Name ethyl 2-hydroxyacetate
SMILES CCOC(=O)CO

Specifications

Color Colorless
Boiling Point 160°C
Quantity 25 g
Formula Weight 104.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Glycolate
RTECSNumber MC5410000
TSCA Yes