missing translation for 'onlineSavingsMsg'
Learn More

Ethyl glycolate, 95%

CAS: 623-50-7 | C4H8O3 | 104.11 g/mol

$201.10 - $201.10

Chemical Identifiers

CAS 623-50-7
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
MDL Number MFCD00021970
InChI Key ZANNOFHADGWOLI-UHFFFAOYSA-N
Synonym ethyl glycolate, ethyl hydroxyacetate, acetic acid, hydroxy-, ethyl ester, glycolic acid, ethyl ester, ethyl glycollate, hydroxyacetic acid ethyl ester, glycolic acid ethyl ester, unii-c306e91wfk, acetic acid, 2-hydroxy-, ethyl ester, 2-hydroxyacetic acid ethyl ester
PubChem CID 12184
IUPAC Name ethyl 2-hydroxyacetate
SMILES CCOC(=O)CO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC410080250
SDS View Documents
Thermo Scientific Chemicals
410080250
25 g Glass bottle
Each for $201.10
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

623-50-7
104.11
ZANNOFHADGWOLI-UHFFFAOYSA-N
12184
CCOC(=O)CO
C4H8O3
MFCD00021970
ethyl glycolate, ethyl hydroxyacetate, acetic acid, hydroxy-, ethyl ester, glycolic acid, ethyl ester, ethyl glycollate, hydroxyacetic acid ethyl ester, glycolic acid ethyl ester, unii-c306e91wfk, acetic acid, 2-hydroxy-, ethyl ester, 2-hydroxyacetic acid ethyl ester
ethyl 2-hydroxyacetate

Specifications

623-50-7
1.0800g/mL
62°C
95%
C4H8O3
HOCH2COOCH2CH3
25 g
ethyl glycolate, ethyl hydroxyacetate, acetic acid, hydroxy-, ethyl ester, glycolic acid, ethyl ester, ethyl glycollate, hydroxyacetic acid ethyl ester, glycolic acid ethyl ester, unii-c306e91wfk, acetic acid, 2-hydroxy-, ethyl ester, 2-hydroxyacetic acid ethyl ester
CCOC(=O)CO
104.11
104.11
Liquid
Colorless
158.0°C to 159.0°C
Authentic
Glass bottle
1.4170 to 1.4200
MFCD00021970
1.08
ZANNOFHADGWOLI-UHFFFAOYSA-N
ethyl 2-hydroxyacetate
12184
95%
Ethyl glycolate
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 210-798-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only