missing translation for 'onlineSavingsMsg'
Learn More

Ethyl glycolate, 95%

Catalog No. AA4147518
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4147518 50 g
AA4147509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4147518 Supplier Thermo Scientific Chemicals Supplier No. 04147518

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 623-50-7 | C4H8O3 | 104.11 g/mol

Ethyl glycolate, is an important building block for the preparation of agrochemicals and medicinal compounds. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff. It is also used in synthetic chemistry, discovery process and R&D. It also can be used as solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 623-50-7
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
MDL Number MFCD00021970
InChI Key ZANNOFHADGWOLI-UHFFFAOYSA-N
Synonym ethyl glycolate, ethyl hydroxyacetate, acetic acid, hydroxy-, ethyl ester, glycolic acid, ethyl ester, ethyl glycollate, hydroxyacetic acid ethyl ester, glycolic acid ethyl ester, unii-c306e91wfk, acetic acid, 2-hydroxy-, ethyl ester, 2-hydroxyacetic acid ethyl ester
PubChem CID 12184
IUPAC Name ethyl 2-hydroxyacetate
SMILES CCOC(=O)CO

Specifications

Color Colorless
Density 1.10 g/mL
Boiling Point 158°C to 159°C
Flash Point 61°C (143°F)
Refractive Index 1.419
Linear Formula HOCH2CO2C2H5
Quantity 50 g
Solubility Information It is very soluble in water,alcohols,acetone,acetates,slightly soluble in ethyl ether,sparingly soluble in hydrocarbon solvents,polymerization occurs above 50 C.
Formula Weight 104.11
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethyl glycolate
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 210-798-4
RTECSNumber MC5410000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only