missing translation for 'onlineSavingsMsg'
Learn More

Ethyl formate, Analytical Standard, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7193384
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018721 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018721 Supplier Merck Emd Millipore Supplier No. 885545MLF
Only null left

Ethyl formate belongs to the group of ethyl esters, which can be synthesized via the reaction between formic acid and ethanol in the presence of sulfuric acid, under thermal conditions.

Specifications

CAS 109-94-4
Vapor Pressure 3.79 psi (20°C)
Vapor Density 2.5 (vs. air)
Density 0.921 g/mL (at 20°C)
Physical Form Neat
Linear Formula HCOOC2H5
MDL Number MFCD00003294
Melting Point -80°C
Boiling Point 52°C to 54°C
Synonym Formic acid ethyl ester
Quantity 5 mL
Formula Weight 74.08
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less