missing translation for 'onlineSavingsMsg'
Learn More

Ethyl fluoroacetate, 98%

CAS: 459-72-3 | C4H7FO2 | 106.096 g/mol

$82.62 - $173.71

Chemical Identifiers

CAS 459-72-3
Molecular Formula C4H7FO2
Molecular Weight (g/mol) 106.096
MDL Number MFCD00000450
InChI Key VCYZVXRKYPKDQB-UHFFFAOYSA-N
Synonym ethyl fluoroacetate, ethyl monofluoroacetate, acetic acid, fluoro-, ethyl ester, fluoroacetic acid ethyl ester, unii-aus1l6um7b, ethylester kyseliny fluoroctove, monofluoroacetic acid ethyl ester, aus1l6um7b, ethylester kyseliny fluoroctove czech, acetic acid, 2-fluoro-, ethyl ester
PubChem CID 9988
IUPAC Name ethyl 2-fluoroacetate
SMILES CCOC(=O)CF
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6134009
View Documents
Thermo Scientific Chemicals
H6134009
10 g
Each for $82.62
Only null left
 
AAH6134018
View Documents
Thermo Scientific Chemicals
H6134018
50 g
Each for $173.71
Only null left
 
Description

Description

Ethyl fluoroacetate is used in the synthesis of 4-deoxy-4-fluoro-muscarines. It is also used as starting reagent for the synthesis of ethyl(diethoxyphosphoryl)fluoroacetate. and in development of an ammonolysis-based microencapsulation process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl fluoroacetate is used in the synthesis of 4-deoxy-4-fluoro-muscarines. It is also used as starting reagent for the synthesis of ethyl(diethoxyphosphoryl)fluoroacetate. and in development of an ammonolysis-based microencapsulation process.

Solubility
Decomposes in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

459-72-3
106.096
VCYZVXRKYPKDQB-UHFFFAOYSA-N
9988
CCOC(=O)CF
C4H7FO2
MFCD00000450
ethyl fluoroacetate, ethyl monofluoroacetate, acetic acid, fluoro-, ethyl ester, fluoroacetic acid ethyl ester, unii-aus1l6um7b, ethylester kyseliny fluoroctove, monofluoroacetic acid ethyl ester, aus1l6um7b, ethylester kyseliny fluoroctove czech, acetic acid, 2-fluoro-, ethyl ester
ethyl 2-fluoroacetate

Specifications

459-72-3
1.093 g/mL
C4H7FO2
FCH2CO2CH2CH3
10 g
1743761
ethyl fluoroacetate, ethyl monofluoroacetate, acetic acid, fluoro-, ethyl ester, fluoroacetic acid ethyl ester, unii-aus1l6um7b, ethylester kyseliny fluoroctove, monofluoroacetic acid ethyl ester, aus1l6um7b, ethylester kyseliny fluoroctove czech, acetic acid, 2-fluoro-, ethyl ester
VCYZVXRKYPKDQB-UHFFFAOYSA-N
ethyl 2-fluoroacetate
9988
98%
Ethyl fluoroacetate
Colorless to Yellow
119°C to 120°C
1.375
MFCD00000450
UN2929
14,4167
Decomposes in water.
CCOC(=O)CF
106.096
106.1
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H226
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Flammable liquid and vapor.

P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P310-P330-P363-P370+P378q-P501c

H226-H300+H310+H330

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.

EINECSNumber : 207-297-8

RTECSNumber : AH7175000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only