missing translation for 'onlineSavingsMsg'
Learn More

Ethyl fluoroacetate, 97%, Thermo Scientific Chemicals

$50.16 - $50.16

Chemical Identifiers

CAS 459-72-3
Molecular Formula C4H7FO2
Molecular Weight (g/mol) 106.1
MDL Number MFCD00000450
InChI Key VCYZVXRKYPKDQB-UHFFFAOYSA-N
Synonym ethyl fluoroacetate, ethyl monofluoroacetate, acetic acid, fluoro-, ethyl ester, fluoroacetic acid ethyl ester, unii-aus1l6um7b, ethylester kyseliny fluoroctove, monofluoroacetic acid ethyl ester, aus1l6um7b, ethylester kyseliny fluoroctove czech, acetic acid, 2-fluoro-, ethyl ester
PubChem CID 9988
IUPAC Name ethyl 2-fluoroacetate
SMILES CCOC(=O)CF
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC172660050
SDS View Documents
Thermo Scientific Chemicals
172660050
5 g Glass bottle
Each for $50.16
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

459-72-3
106.1
VCYZVXRKYPKDQB-UHFFFAOYSA-N
9988
CCOC(=O)CF
C4H7FO2
MFCD00000450
ethyl fluoroacetate, ethyl monofluoroacetate, acetic acid, fluoro-, ethyl ester, fluoroacetic acid ethyl ester, unii-aus1l6um7b, ethylester kyseliny fluoroctove, monofluoroacetic acid ethyl ester, aus1l6um7b, ethylester kyseliny fluoroctove czech, acetic acid, 2-fluoro-, ethyl ester
ethyl 2-fluoroacetate

Specifications

459-72-3
1.0900g/mL
30°C
97%
C4H7FO2
FCH2CO2C2H5
5 g
05,304
15, 4199
Solubility in water: decomposes.
CCOC(=O)CF
106.1
106.1
Liquid
Colorless
119.2°C (753.0 mmHg)
Authentic
Glass bottle
1.3745 to 1.3765
MFCD00000450
02, 193
1.09
ethyl fluoroacetate, ethyl monofluoroacetate, acetic acid, fluoro-, ethyl ester, fluoroacetic acid ethyl ester, unii-aus1l6um7b, ethylester kyseliny fluoroctove, monofluoroacetic acid ethyl ester, aus1l6um7b, ethylester kyseliny fluoroctove czech, acetic acid, 2-fluoro-, ethyl ester
VCYZVXRKYPKDQB-UHFFFAOYSA-N
ethyl 2-fluoroacetate
9988
97%
Ethyl fluoroacetate, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Fatal in contact with skin.
Fatal if inhaled.
Fatal if swallowed.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 207-297-8

RTECSNumber : AH7175000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only