missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Ether, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, contains no preservative, >99.9%, Honeywell™ Burdick & Jackson
Supplier: Honeywell-Burdick and Jackson 1074
Specifications
| Ethyl Ether | |
| 60-29-7 | |
| 100.0000% | |
| 35°C | |
| −45°C | |
| C4H10O | |
| UN1155 | |
| diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
| CCOCC | |
| 74.12 | |
| CHEBI:35702 | |
| >99.9% |
| B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, contains no preservative, >99.9% | |
| 0.08 AU max. at 250 nm (UV), 0.01 AU max. at 275 nm (UV), 0.005 AU max. at 300 nm (UV) | |
| 0.713 g/cm3 | |
| 4 x 4 L | |
| Glass Bottle | |
| MFCD00011646 | |
| 1696894 | |
| RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
| ethoxyethane | |
| 3283 | |
| 74.12 | |
| HPLC |
Chemical Identifiers
| 60-29-7 | |
| 74.12 | |
| RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
| 3283 | |
| ethoxyethane |
| C4H10O | |
| MFCD00011646 | |
| diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
| CHEBI:35702 | |
| CCOCC |
Safety and Handling
ShelfLife : 365 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: I; Proper Shipping Name: Diethyl ether