missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Ether, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, contains no preservative, >99.9%, Honeywell™ Burdick & Jackson

Catalog No. p-7111742
Change view
Click to view available options
Quantity:
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000531 6 x 1 L Glass Bottle
6000529 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000531 Supplier Honeywell Chemicals Supplier No. 1071L
Only null left

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Absorbance 0.08 AU max. at 250 nm (UV), 0.01 AU max. at 275 nm (UV), 0.005 AU max. at 300 nm (UV)
CAS Max % 100.0000%
Density 0.713 g/cm3
Quantity 6 x 1 L
Flash Point −45°C
Packaging Glass Bottle
UN Number UN1155
Beilstein 1696894
Boiling Point 35°C
Formula Weight 74.12
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material Ethyl Ether, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, contains no preservative, >99.9%
Show More Show Less
ShelfLife 365 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: I; Proper Shipping Name: Diethyl ether