missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Ether, ACS, 98%, Spectrum™ Chemical

Catalog No. 18610430
Change view
Click to view available options
Quantity:
4 L
Packaging:
Steel Can
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610430 4 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18610430 Supplier Spectrum Chemical Mfg Cor Supplier No. E10154LTMT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 60-29-7 , 64-17-5 , 128-37-0

Specifications

CAS Max % 100%, <3%, <0.1%
Quantity 4 L
Packaging Steel Can
Residue after Evaporation 1e-05
Titratable Acid 0.0002 meq/g
Boiling Point 34°C
Percent Purity 0.98
Grade Reagent
Chemical Name or Material Ethyl Ether

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.