missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Erucate 95.0+%, TCI America™
$132.84 - $450.49
Chemical Identifiers
| CAS | 37910-77-3 |
|---|---|
| Molecular Formula | C24H46O2 |
| Molecular Weight (g/mol) | 366.63 |
| MDL Number | MFCD00027405 |
| InChI Key | WFZQLUSOXHIVKL-QXMHVHEDSA-N |
| Synonym | cis-13-Docosenoic Acid Ethyl Ester, Ethyl cis-13-Docosenoate, Erucic Acid Ethyl Ester |
| PubChem CID | 5364508 |
| ChEBI | CHEBI:84891 |
| IUPAC Name | ethyl (13Z)-docos-13-enoate |
| SMILES | CCCCCCCC\C=C/CCCCCCCCCCCC(=O)OCC |
Chemical Identifiers
| 37910-77-3 | |
| 366.63 | |
| WFZQLUSOXHIVKL-QXMHVHEDSA-N | |
| 5364508 | |
| ethyl (13Z)-docos-13-enoate |
| C24H46O2 | |
| MFCD00027405 | |
| cis-13-Docosenoic Acid Ethyl Ester, Ethyl cis-13-Docosenoate, Erucic Acid Ethyl Ester | |
| CHEBI:84891 | |
| CCCCCCCC\C=C/CCCCCCCCCCCC(=O)OCC |
Specifications
| 37910-77-3 | |
| 233°C | |
| MFCD00027405 | |
| cis-13-Docosenoic Acid Ethyl Ester, Ethyl cis-13-Docosenoate, Erucic Acid Ethyl Ester | |
| CCCCCCCC\C=C/CCCCCCCCCCCC(=O)OCC | |
| 366.63 | |
| CHEBI:84891 | |
| ≥95.0% (GC) | |
| Ethyl Erucate |
| Yellow | |
| C24H46O2 | |
| 5 mL | |
| WFZQLUSOXHIVKL-QXMHVHEDSA-N | |
| ethyl (13Z)-docos-13-enoate | |
| 5364508 | |
| 366.63 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0798
TSCA : No