accessibility menu, dialog, popup

UserName

CAS RN 37910-77-3

H3COOCH3(Z)

CAS RN 37910-77-3

IUPAC Name: ethyl (13Z)-docos-13-enoate
Synonyms: ethyl (13Z)-docos-13-enoate
Molecular Weight (g/mol): 366.63
Molecular Formula: C24H46O2
InChi Key: WFZQLUSOXHIVKL-QXMHVHEDSA-N
SMILES: CCCCCCCC\C=C/CCCCCCCCCCCC(=O)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results