missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Erucate 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 233°C |
| Quantity | 5 mL |
| Formula Weight | 366.63 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Ethyl Erucate |
Safety and Handling
| EINECSNumber | (2)-0798 |
| TSCA | No |