missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (E)-3-(1-pyrrolidinyl)crotonate, 97%

Catalog No. p-7043366
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
AAL0046106 5 g
AAL0046114 25 g
2 options

Catalog No. AAL0046106

Supplier: Thermo Scientific Chemicals L0046106

Only null left

CAS: 54716-02-8 | C10H17NO2 | 183.251 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54716-02-8
Molecular Formula C10H17NO2
Molecular Weight (g/mol) 183.251
MDL Number MFCD00014097
InChI Key MSOQKPXSIHLODG-CMDGGOBGSA-N
Synonym e-ethyl 3-pyrrolidin-1-yl but-2-enoate, ethyl e-3-1-pyrrolidinyl crotonate, 2e-3-1-pyrrolidinyl-2-butenoic acid ethyl ester, ethyl e-3-1-pyrrolidino crotonate, ethyl e-3-pyrrolidin-1-ylbut-2-enoate, e-3-pyrrolidin-1-yl-but-2-enoic acid ethyl ester, 2-butenoic acid, 3-1-pyrrolidinyl-, ethyl ester, e, ethyl 2e-3-pyrrolidin-1-yl but-2-enoate, ethyl 3-1-pyrrolidinyl crotonate
PubChem CID 736203
IUPAC Name ethyl (E)-3-pyrrolidin-1-ylbut-2-enoate
SMILES CCOC(=O)C=C(C)N1CCCC1

Specifications

Boiling Point 125°C to 126°C (0.8 mmHg)
Refractive Index 1.541
Quantity 5 g
Beilstein 1310113
Formula Weight 183.25
Percent Purity 97%
Chemical Name or Material Ethyl (E)-3-(1-pyrrolidinyl)crotonate
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 259-303-3
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only