missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl dodecanoate, 98%
CAS: 106-33-2 | C14H28O2 | 228.38 g/mol
Supplier: Thermo Scientific Chemicals A111880F
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Ethyl dodecanoate | |
| -2°C | |
| 268°C to 270°C | |
| Fruit-like | |
| 1.4315 | |
| 2500 mL | |
| 14,3818 | |
| MMXKVMNBHPAILY-UHFFFAOYSA-N | |
| ethyl dodecanoate | |
| 7800 | |
| 228.38 |
| 106-33-2 | |
| 0.862 | |
| 138°C (280°F) | |
| C14H28O2 | |
| MFCD00015065 | |
| 1769671 | |
| ethyl laurate, dodecanoic acid, ethyl ester, ethyl laurinate, ethyl dodecylate, lauric acid ethyl ester, lauric acid, ethyl ester, ethyl laurate natural, dodecanoic acid ethyl ester, ethyl n-dodecanote, unii-f389d4md5k | |
| CCCCCCCCCCCC(=O)OCC | |
| 228.38 | |
| CHEBI:87427 | |
| 98% |
Chemical Identifiers
| 106-33-2 | |
| 228.38 | |
| MMXKVMNBHPAILY-UHFFFAOYSA-N | |
| 7800 | |
| ethyl dodecanoate |
| C14H28O2 | |
| MFCD00015065 | |
| ethyl laurate, dodecanoic acid, ethyl ester, ethyl laurinate, ethyl dodecylate, lauric acid ethyl ester, lauric acid, ethyl ester, ethyl laurate natural, dodecanoic acid ethyl ester, ethyl n-dodecanote, unii-f389d4md5k | |
| CHEBI:87427 | |
| CCCCCCCCCCCC(=O)OCC |
Safety and Handling
EINECSNumber : 203-386-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only