missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl DL-3-Hydroxybutyrate 98.0+%, TCI America™
$55.84 - $329.20
Chemical Identifiers
| CAS | 5405-41-4 |
|---|---|
| Molecular Formula | C6H12O3 |
| Molecular Weight (g/mol) | 132.16 |
| MDL Number | MFCD00004545 |
| InChI Key | OMSUIQOIVADKIM-UHFFFAOYNA-N |
| Synonym | ethyl 3-hydroxybutyrate, ethyl beta-hydroxybutyrate, butanoic acid, 3-hydroxy-, ethyl ester, butyric acid, 3-hydroxy-, ethyl ester, fema no. 3428, ethyl 3-hydroxybutyrate natural, ethyl dl-3-hydroxybutyrate, ethyl 1-3-hydroxybutyrate, dl-3-hydroxy-n-butyric acid ethyl ester, ethyl3-hydroxybutyrate |
| PubChem CID | 62572 |
| ChEBI | CHEBI:87685 |
| IUPAC Name | ethyl 3-hydroxybutanoate |
| SMILES | CCOC(=O)CC(C)O |
Chemical Identifiers
| 5405-41-4 | |
| 132.16 | |
| OMSUIQOIVADKIM-UHFFFAOYNA-N | |
| 62572 | |
| ethyl 3-hydroxybutanoate |
| C6H12O3 | |
| MFCD00004545 | |
| ethyl 3-hydroxybutyrate, ethyl beta-hydroxybutyrate, butanoic acid, 3-hydroxy-, ethyl ester, butyric acid, 3-hydroxy-, ethyl ester, fema no. 3428, ethyl 3-hydroxybutyrate natural, ethyl dl-3-hydroxybutyrate, ethyl 1-3-hydroxybutyrate, dl-3-hydroxy-n-butyric acid ethyl ester, ethyl3-hydroxybutyrate | |
| CHEBI:87685 | |
| CCOC(=O)CC(C)O |
Specifications
| 5405-41-4 | |
| 180°C | |
| MFCD00004545 | |
| ethyl 3-hydroxybutyrate, ethyl beta-hydroxybutyrate, butanoic acid, 3-hydroxy-, ethyl ester, butyric acid, 3-hydroxy-, ethyl ester, fema no. 3428, ethyl 3-hydroxybutyrate natural, ethyl dl-3-hydroxybutyrate, ethyl 1-3-hydroxybutyrate, dl-3-hydroxy-n-butyric acid ethyl ester, ethyl3-hydroxybutyrate | |
| CCOC(=O)CC(C)O | |
| 132.16 | |
| CHEBI:87685 | |
| ≥98.0% (GC) | |
| Ethyl DL-3-Hydroxybutyrate |
| Colorless | |
| C6H12O3 | |
| 25 g | |
| OMSUIQOIVADKIM-UHFFFAOYNA-N | |
| ethyl 3-hydroxybutanoate | |
| 62572 | |
| 132.16 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-1430
TSCA : Yes