missing translation for 'onlineSavingsMsg'
Learn More

Ethyl DL-3-Hydroxybutyrate 98.0+%, TCI America™

Catalog No. H0230500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0230500G 500 g
H023025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0230500G Supplier TCI America Supplier No. H0230500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5405-41-4
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00004545
InChI Key OMSUIQOIVADKIM-UHFFFAOYNA-N
Synonym ethyl 3-hydroxybutyrate, ethyl beta-hydroxybutyrate, butanoic acid, 3-hydroxy-, ethyl ester, butyric acid, 3-hydroxy-, ethyl ester, fema no. 3428, ethyl 3-hydroxybutyrate natural, ethyl dl-3-hydroxybutyrate, ethyl 1-3-hydroxybutyrate, dl-3-hydroxy-n-butyric acid ethyl ester, ethyl3-hydroxybutyrate
PubChem CID 62572
ChEBI CHEBI:87685
IUPAC Name ethyl 3-hydroxybutanoate
SMILES CCOC(=O)CC(C)O

Specifications

Color Colorless
Boiling Point 180°C
Quantity 500 g
Formula Weight 132.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl DL-3-Hydroxybutyrate
EINECSNumber (2)-1430
TSCA Yes