missing translation for 'onlineSavingsMsg'
Learn More

Ethyl(dimethyl)(2-phenylethyl)ammonium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™

Catalog No. p-7129423
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12831G 1 g
E12835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E12831G Supplier TCI America Supplier No. E12831G
Only null left

Chemical Identifiers

CAS 1804970-28-2
Molecular Formula C14H20F6N2O4S2
Molecular Weight (g/mol) 458.434
InChI Key XVNMSYLRWLNKMT-UHFFFAOYSA-N
Synonym Ethyl(dimethyl)phenethylammonium Bis(trifluoromethanesulfonyl)imide
PubChem CID 121234255
IUPAC Name bis(trifluoromethylsulfonyl)azanide;ethyl-dimethyl-(2-phenylethyl)azanium
SMILES CC[N+](C)(C)CCC1=CC=CC=C1.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Quantity 1 g
Formula Weight 458.43
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Ethyl(dimethyl)(2-phenylethyl)ammonium Bis(trifluoromethanesulfonyl)imide