missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Diethoxyacetate 97.0+%, TCI America™
Supplier: TCI America D1883100ML
Specifications
| Ethyl Diethoxyacetate | |
| Colorless | |
| C8H16O4 | |
| 100 mL | |
| XCLBIKIQSCTANZ-UHFFFAOYSA-N | |
| ethyl 2,2-diethoxyacetate | |
| 80169 | |
| ≥97.0% (GC) |
| 6065-82-3 | |
| 84°C | |
| MFCD00009231 | |
| ethyl diethoxyacetate, acetic acid, diethoxy-, ethyl ester, diethoxyacetic acid ethyl ester, ethyldiethoxyacetate, acetic acid, 2,2-diethoxy-, ethyl ester, acetic acid,diethoxy,ethyl ester, acmc-209mlb, ethyl bis ethyloxy acetate, ethyl glyoxylate diethyl acetal | |
| CCOC(C(=O)OCC)OCC | |
| 176.212 | |
| 176.21 | |
| Liquid |
Chemical Identifiers
| 6065-82-3 | |
| 176.212 | |
| XCLBIKIQSCTANZ-UHFFFAOYSA-N | |
| 80169 | |
| CCOC(C(=O)OCC)OCC |
| C8H16O4 | |
| MFCD00009231 | |
| ethyl diethoxyacetate, acetic acid, diethoxy-, ethyl ester, diethoxyacetic acid ethyl ester, ethyldiethoxyacetate, acetic acid, 2,2-diethoxy-, ethyl ester, acetic acid,diethoxy,ethyl ester, acmc-209mlb, ethyl bis ethyloxy acetate, ethyl glyoxylate diethyl acetal | |
| ethyl 2,2-diethoxyacetate |
Safety and Handling
TSCA : Yes