missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Diethoxyacetate 97.0+%, TCI America™

Catalog No. p-7126990
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D188325ML 25 mL
D1883100ML 100 mL
D1883500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D188325ML Supplier TCI America Supplier No. D188325ML
Only null left

Chemical Identifiers

CAS 6065-82-3
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.212
MDL Number MFCD00009231
InChI Key XCLBIKIQSCTANZ-UHFFFAOYSA-N
Synonym ethyl diethoxyacetate, acetic acid, diethoxy-, ethyl ester, diethoxyacetic acid ethyl ester, ethyldiethoxyacetate, acetic acid, 2,2-diethoxy-, ethyl ester, acetic acid,diethoxy,ethyl ester, acmc-209mlb, ethyl bis ethyloxy acetate, ethyl glyoxylate diethyl acetal
PubChem CID 80169
IUPAC Name ethyl 2,2-diethoxyacetate
SMILES CCOC(C(=O)OCC)OCC

Specifications

Color Colorless
Boiling Point 84°C
Quantity 25 mL
Formula Weight 176.21
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Diethoxyacetate