missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Di-o-tolylphosphonoacetate 95.0+%, TCI America™

Catalog No. D25481G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25481G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D25481G Supplier TCI America Supplier No. D25481G

Chemical Identifiers

CAS 188945-41-7
Molecular Formula C18H21O5P
Molecular Weight (g/mol) 348.335
MDL Number MFCD01321166
InChI Key BOEGWWFFVZDOIH-UHFFFAOYSA-N
Synonym Di-o-tolylphosphonoacetic Acid Ethyl Ester
PubChem CID 10593815
IUPAC Name ethyl 2-bis(2-methylphenoxy)phosphorylacetate
SMILES CCOC(=O)CP(=O)(OC1=CC=CC=C1C)OC2=CC=CC=C2C

Specifications

Boiling Point 240°C
Quantity 1 g
Formula Weight 348.33
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Di-o-tolylphosphonoacetate [Horner-Emmons Reagent]