missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Cyanoacetate 99.0+%, TCI America™

Catalog No. p-7124422
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C044125G 25 g
C0441500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C044125G Supplier TCI America Supplier No. C044125G
Only null left

Chemical Identifiers

CAS 105-56-6
Molecular Formula C5H7NO2
Molecular Weight (g/mol) 113.12
MDL Number MFCD00001940
InChI Key ZIUSEGSNTOUIPT-UHFFFAOYSA-N
Synonym ethyl cyanoacetate, ethyl cyanacetate, cyanoacetic acid ethyl ester, acetic acid, cyano-, ethyl ester, ethyl cyanoethanoate, cyanoacetic ester, cyanacetate ethyle, usaf kf-25, estere cianoacetico, ethyl cyano acetate
PubChem CID 7764
IUPAC Name ethyl 2-cyanoacetate
SMILES CCOC(=O)CC#N

Specifications

Melting Point -22°C
Color Colorless
Boiling Point 206°C
Quantity 25 g
Formula Weight 113.12
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Cyanoacetate
EINECSNumber (2)-1549
RTECSNumber AG4110000
TSCA Yes