missing translation for 'onlineSavingsMsg'
Learn More

Ethyl cyanoacetate, 98+%

Catalog No. p-7081494
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1149836 500 g
AAA114980E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1149836 Supplier Thermo Scientific Chemicals Supplier No. A1149836

CAS: 105-56-6 | C5H7NO2 | 113.12 g/mol

Cyanoacetate is a building block for the synthesis of heterocycles and are are used as drugs. It is also used as Pharmaceutical intermediates, or Oil soluble couplers of film raw material . It is also used as a reagent in labelled pyrimidine and purine synthesis. It is used as an intermediate of fine chemicals such as medicine and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyanoacetate is a building block for the synthesis of heterocycles and are are used as drugs. It is also used as Pharmaceutical intermediates, or Oil soluble couplers of film raw material . It is also used as a reagent in labelled pyrimidine and purine synthesis. It is used as an intermediate of fine chemicals such as medicine and dyestuff.

Solubility
It is slightly soluble in water. Mostly soluble in organic solvents such as chloroform, ethyl acetate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials strong acids, strong bases, strong oxidizing agents, strong reducing agents.

Chemical Identifiers

CAS 105-56-6
Molecular Formula C5H7NO2
Molecular Weight (g/mol) 113.12
MDL Number MFCD00001940
InChI Key ZIUSEGSNTOUIPT-UHFFFAOYSA-N
Synonym ethyl cyanoacetate, ethyl cyanacetate, cyanoacetic acid ethyl ester, acetic acid, cyano-, ethyl ester, ethyl cyanoethanoate, cyanoacetic ester, cyanacetate ethyle, usaf kf-25, estere cianoacetico, ethyl cyano acetate
PubChem CID 7764
IUPAC Name ethyl 2-cyanoacetate
SMILES CCOC(=O)CC#N

Specifications

Melting Point -23°C
Density 1.063
Boiling Point 208°C to 210°C
Flash Point 109°C (228°F)
Odor Weak
Refractive Index 1.418
Linear Formula NCCH2CO2CH2CH3
Quantity 500 g
Beilstein 605871
Merck Index 14,3786
Solubility Information It is slightly soluble in water. Mostly soluble in organic solvents such as chloroform,ethyl acetate.
Formula Weight 113.12
Percent Purity ≥98%
Chemical Name or Material Ethyl cyanoacetate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 203-309-0
RTECSNumber AG4110000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only