missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Crotonate 98.0+%, TCI America™

Catalog No. C041825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C041825ML 25 mL
C0418500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C041825ML Supplier TCI America Supplier No. C041825ML

Chemical Identifiers

CAS 623-70-1
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00009289
InChI Key ZFDIRQKJPRINOQ-HWKANZROSA-N
Synonym ethyl crotonate, ethyl trans-crotonate, ethyl e-but-2-enoate, ethyl trans-2-butenoate, crotonic acid, ethyl ester, 2-butenoic acid, ethyl ester, 2e, ethyl e-2-butenoate, 2-butenoic acid, ethyl ester, ethyl 2e-but-2-enoate, crotonic acid, ethyl ester, e
PubChem CID 429065
IUPAC Name ethyl (2E)-but-2-enoate
SMILES CCOC(=O)\C=C\C

Specifications

Color Colorless
Boiling Point 138°C
Quantity 25 mL
UN Number 1862
Formula Weight 114.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Crotonate
EINECSNumber (2)-0966
RTECSNumber GQ3500000
TSCA Yes