missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl crotonate, 98%
Description
Ethyl crotonate is used as a solvent for cellulose esters as well as a plasticizer for acrylic resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl crotonate is used as a solvent for cellulose esters as well as a plasticizer for acrylic resins.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Ethyl crotonate is used as a solvent for cellulose esters as well as a plasticizer for acrylic resins.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
| Density | 0.918 |
| Boiling Point | 141°C to 143°C |
| Flash Point | 2°C (36°F) |
| Quantity | 100 g |
| UN Number | UN1862 |
| Beilstein | 635834 |
| Solubility Information | Not miscible in water. |
| Formula Weight | 114.14 |
| Percent Purity | 98% |
| Chemical Name or Material | Ethyl crotonate |
Safety and Handling
| Hazard Category | H225-H319 |
| Hazard Statement | GHS H Statement H225-H314-H318 Highly flammable liquid and vapour. Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYL CROTONATE |
| EINECSNumber | 210-808-7 |
| RTECSNumber | GQ3500000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only