missing translation for 'onlineSavingsMsg'
Learn More

Ethyl crotonate, 98%

Catalog No. AAA1252936
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1252936 500 g
AAA1252922 100 g
AAA125290E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1252936 Supplier Thermo Scientific Chemicals Supplier No. A1252936
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 623-70-1 | C6H10O2 | 114.14 g/mol

Ethyl crotonate is used as a solvent for cellulose esters as well as a plasticizer for acrylic resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl crotonate is used as a solvent for cellulose esters as well as a plasticizer for acrylic resins.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 623-70-1
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00009289
InChI Key ZFDIRQKJPRINOQ-HWKANZROSA-N
Synonym ethyl crotonate, ethyl trans-crotonate, ethyl e-but-2-enoate, ethyl trans-2-butenoate, crotonic acid, ethyl ester, 2-butenoic acid, ethyl ester, 2e, ethyl e-2-butenoate, 2-butenoic acid, ethyl ester, ethyl 2e-but-2-enoate, crotonic acid, ethyl ester, e
PubChem CID 429065
IUPAC Name ethyl (E)-but-2-enoate
SMILES CCOC(=O)\C=C\C

Specifications

Density 0.918
Boiling Point 141°C to 143°C
Flash Point 2°C (36°F)
Quantity 500 g
UN Number UN1862
Beilstein 635834
Solubility Information Not miscible in water.
Formula Weight 114.14
Percent Purity 98%
Chemical Name or Material Ethyl crotonate
Hazard Category H225-H319
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYL CROTONATE
EINECSNumber 210-808-7
RTECSNumber GQ3500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only