missing translation for 'onlineSavingsMsg'
Learn More

Ethyl coumarin-3-carboxylate, 98%

Catalog No. AAA1255306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1255306 5 g
AAA1255303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1255306 Supplier Thermo Scientific Chemicals Supplier No. A1255306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1846-76-0 | C12H10O4 | 218.208 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1846-76-0
Molecular Formula C12H10O4
Molecular Weight (g/mol) 218.208
MDL Number MFCD00016964
InChI Key XKHPEMKBJGUYCM-UHFFFAOYSA-N
Synonym ethyl coumarin-3-carboxylate, ethyl 2-oxo-2h-chromene-3-carboxylate, 3-ethoxycarbonylcoumarin, ethyl 3-coumarincarboxylate, 3-carbethoxycoumarin, ethylcoumarin-3-carboxylate, coumarin-3-carboxylic acid ethyl ester, 1,2-benzopyran-2-one-3-carboxylic acid, ethyl ester, 2h-1-benzopyran-3-carboxylic acid, 2-oxo-, ethyl ester, 2h-1-benzopyran-3-carboxylicacid, 2-oxo-, ethyl ester
PubChem CID 15800
IUPAC Name ethyl 2-oxochromene-3-carboxylate
SMILES CCOC(=O)C1=CC2=CC=CC=C2OC1=O

Specifications

Melting Point 92°C to 95°C
Quantity 5 g
Beilstein 200147
Formula Weight 218.21
Percent Purity 98%
Chemical Name or Material Ethyl coumarin-3-carboxylate
RTECSNumber DJ2501000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only