accessibility menu, dialog, popup

UserName

CAS RN 1846-76-0

IUPAC Name: ethyl 2-oxo-2H-chromene-3-carboxylate
Synonyms: ethyl 2-oxochromene-3-carboxylate
Molecular Weight (g/mol): 218.21
Molecular Formula: C12H10O4
InChi Key: XKHPEMKBJGUYCM-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC2=CC=CC=C2OC1=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)