missing translation for 'onlineSavingsMsg'
Learn More

Ethyl cis-9-Hexadecenoate 95.0+%, TCI America™

Supplier:  TCI America H0501100MG

 View more versions of this product

Catalog No. H0501100MG

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethyl cis-9-Hexadecenoate
Colorless
C18H34O2
100 mg
JELGPLUONQGOHF-KTKRTIGZSA-N
ethyl (Z)-hexadec-9-enoate
6436624
282.47
Liquid
56219-10-4
135°C
MFCD00056314
Ethyl Palmitoleate, cis-9-Hexadecenoic Acid Ethyl Ester, Palmitoleic Acid Ethyl Ester
CCCCCCC=CCCCCCCCC(=O)OCC
282.468
CHEBI:84934
≥95.0% (GC)

Chemical Identifiers

56219-10-4
282.468
JELGPLUONQGOHF-KTKRTIGZSA-N
6436624
ethyl (Z)-hexadec-9-enoate
C18H34O2
MFCD00056314
Ethyl Palmitoleate, cis-9-Hexadecenoic Acid Ethyl Ester, Palmitoleic Acid Ethyl Ester
CHEBI:84934
CCCCCCC=CCCCCCCCC(=O)OCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions