CAS RN 56219-10-4
CAS RN 56219-10-4
IUPAC Name:
ethyl (9Z)-hexadec-9-enoate
Synonyms:
ethyl (9Z)-hexadec-9-enoate
Molecular Weight (g/mol):
282.47
Molecular Formula:
C18H34O2
InChi Key:
JELGPLUONQGOHF-KTKRTIGZSA-N
SMILES:
CCCCCC\C=C/CCCCCCCC(=O)OCC
1
–
1
of
1
results
1
Ethyl cis-9-Hexadecenoate 95.0+%, TCI America™
CAS: 56219-10-4 Molecular Formula: C18H34O2 Molecular Weight (g/mol): 282.468 MDL Number: MFCD00056314 InChI Key: JELGPLUONQGOHF-KTKRTIGZSA-N Synonym: Ethyl Palmitoleate, cis-9-Hexadecenoic Acid Ethyl Ester, Palmitoleic Acid Ethyl Ester PubChem CID: 6436624 ChEBI: CHEBI:84934 IUPAC Name: ethyl (Z)-hexadec-9-enoate SMILES: CCCCCCC=CCCCCCCCC(=O)OCC