missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Cinnamate 99.0+%, TCI America™

Catalog No. p-7124370
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C035925G 25 g
C0359100G 100 g
C0359500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C035925G Supplier TCI America Supplier No. C035925G
Only null left

Chemical Identifiers

CAS 103-36-6
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD00009189
InChI Key KBEBGUQPQBELIU-CMDGGOBGSA-N
Synonym ethyl cinnamate, ethyl 3-phenylacrylate, ethylcinnamate, cinnamic acid ethyl ester, ethylcinnamoate, cinnamic acid, ethyl ester, ethyl trans-cinnamate, e-ethyl cinnamate, ethyl 3-phenyl-2-propenoate, ethyl benzylideneacetate
PubChem CID 637758
ChEBI CHEBI:4895
IUPAC Name ethyl (E)-3-phenylprop-2-enoate
SMILES CCOC(=O)C=CC1=CC=CC=C1

Specifications

Melting Point 10°C
Color Yellow
Boiling Point 271°C
Quantity 25 g
Formula Weight 176.22
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Cinnamate
EINECSNumber (3)-1750
RTECSNumber GD9010000
TSCA Yes