missing translation for 'onlineSavingsMsg'
Learn More

Ethyl cinnamate, 98+%

Catalog No. p-7020824
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1290622 100 g
AAA1290636 500 g
AAA129060E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1290622 Supplier Thermo Scientific Chemicals Supplier No. A1290622

CAS: 103-36-6 | C11H12O2 | 176.215 g/mol

Ethyl cinnamate is used in medical intermediate, as a pharmaceutical intermediate and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl cinnamate is used in medical intermediate, as a pharmaceutical intermediate and in organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, acids, bases and reducing agents.

Chemical Identifiers

CAS 103-36-6
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD00009189
InChI Key KBEBGUQPQBELIU-CMDGGOBGSA-N
Synonym ethyl cinnamate, ethyl 3-phenylacrylate, ethylcinnamate, cinnamic acid ethyl ester, ethylcinnamoate, cinnamic acid, ethyl ester, ethyl trans-cinnamate, e-ethyl cinnamate, ethyl 3-phenyl-2-propenoate, ethyl benzylideneacetate
PubChem CID 637758
ChEBI CHEBI:4895
IUPAC Name ethyl (E)-3-phenylprop-2-enoate
SMILES CCOC(=O)C=CC1=CC=CC=C1

Specifications

Melting Point 7°C
Density 1.049
Boiling Point 270°C to 272°C
Flash Point 135°C (275°F)
Assay Percent Range ≥98%
Refractive Index 1.559
Quantity 100 g
Beilstein 1238804
Merck Index 14,2299
Solubility Information Insoluble in water.
Formula Weight 176.22
Percent Purity ≥98%
Chemical Name or Material Ethyl cinnamate
Show More Show Less
EINECSNumber 203-104-6
RTECSNumber GD9010000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only