missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Chlorodifluoroacetate 97.0+%, TCI America™

Catalog No. p-7124996
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
C145825G 25 g
C1458500G 500 g
2 options

Catalog No. C145825G

Supplier: TCI America C145825G

Only null left

Chemical Identifiers

CAS 383-62-0
Molecular Formula C4H5ClF2O2
Molecular Weight (g/mol) 158.529
MDL Number MFCD00013662
InChI Key GVCAWQUJCHZRCB-UHFFFAOYSA-N
Synonym ethyl chlorodifluoroacetate, chlorodifluoroacetic acid ethyl ester, ethyl chlorodifluoroethanoate, acetic acid, chlorodifluoro-, ethyl ester, ethylchlorodifluoroacetate, acetic acid, 2-chloro-2,2-difluoro-, ethyl ester, ethyl 2-chloro-2,2-difluoro-acetate, pubchem12573, ethyl chlordifluoroacetate, acmc-1cr8n
PubChem CID 67843
IUPAC Name ethyl 2-chloro-2,2-difluoroacetate
SMILES CCOC(=O)C(F)(F)Cl

Specifications

Color Colorless
Boiling Point 96°C
Quantity 25 g
UN Number 2924
Formula Weight 158.53
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Chlorodifluoroacetate